Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5001P

Sigma-Aldrich

Bovine Serum Albumin(BSA) Sampling Kit

≥98%, USA origin, enzyme immunoassay: suitable, ELISA: suitable, Serology: suitable

Synonym(s):

Lyophilized Powder Sampling Kit

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41141801
NACRES:
NA.28

Product Name

Bovine Serum Albumin(BSA) Sampling Kit, lyophilized powder

form

lyophilized powder

Quality Level

packaging

pkg of 4 × 10 g

origin

USA origin

technique(s)

ELISA: suitable
enzyme immunoassay: suitable using

dilution

(Serology)

foreign activity

Bluetongue, none detected
Vesicular Stomatitis Virus, none detected

Looking for similar products? Visit Product Comparison Guide

Components

81003N, 81066N, 820453, 821007

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service