Skip to Content
MilliporeSigma

5.00111

SAFC

Cocarboxylase tetrahydrate

(thiamine pyrophosphate tetrahydrate), EMPROVE® API

Pharma Manufacturing

Synonym(s):

Cocarboxylase tetrahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18N4O7P2S · 4H2O
CAS Number:
Molecular Weight:
496.37
UNSPSC Code:
12352107
EC Index Number:
205-230-7

Quality Level

product line

EMPROVE® API

form

solid

pH

2.4-3.2 (20 °C, 100 g/L in H2O)

solubility

55 g/L

bulk density

450 kg/m3

application(s)

liquid formulation
pharmaceutical
semi-solid formulation
solid formulation

storage temp.

2-25°C

InChI

1S/C12H18N4O7P2S.4H2O/c1-8-11(3-4-22-25(20,21)23-24(17,18)19)26-7-16(8)6-10-5-14-9(2)15-12(10)13;;;;/h5,7H,3-4,6H2,1-2H3,(H4-,13,14,15,17,18,19,20,21);4*1H2

InChI key

KLOLNRGPRQYTIW-UHFFFAOYSA-N

General description

We provide industry-leading production know-how and comprehensive regulatory support for high-quality APIs, backed by nearly 200 years of experience. From a complete portfolio of innovative folates to custom synthesis, our customized and scalable solutions meet your specific production needs. As part of our EMPROVE® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Cocarboxylase tetrahydrate Emprove® API is a source for Vitamin B1 for use in clinical nutrition, vitamin supplements and infusion for Vitamin B1 deficiency. Cocarboxylase Tetrahydrate (thiamine diphosphoric acid ester tetrahydrate) Emprove® API is manufactured according to ICH Q7 guidelines. An ASMF is available.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

The substance 5.00111 has no REACH registration because the only supported use is the use in medicinal products for human or veterinary use or in food or feedingstuffs according to article 2 of the REACH Regulation (EC) No 1907/2006.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ferenc Tóth et al.
Bone, 132, 115214-115214 (2019-12-31)
Bone morphogenetic protein 2 (BMP-2) is a member of the transforming growth factor-β superfamily, it is known to be a factor involved in skeletal development and capable of inducing in vitro osteogenic differentiation of mesenchymal stem cells (MSCs). Dental pulp

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service