Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

500105

Millipore

Oxytetracycline, Hydrochloride

An antimicrobial agent. A member of the tetracycline family that inhibits bacterial protein synthesis by binding to the 30S ribosomal subunit.

Synonym(s):

Oxytetracycline, Hydrochloride, OTC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H24N2O9 · xHCl
CAS Number:
Molecular Weight:
460.43 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥95% (HPLC)

form

solid

optical activity

[α]/D -190 to -200°, c = 1 in 0.1 M HCl (anhydrous basis)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

color

yellow

solubility

water: soluble

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C22H24N2O9.ClH/c1-21(32)7-5-4-6-8(25)9(7)15(26)10-12(21)17(28)13-14(24(2)3)16(27)11(20(23)31)19(30)22(13,33)18(10)29;/h4-6,12-14,17,25-26,28,31-33H,23H2,1-3H3;1H/b20-11-;/t12-,13-,14+,17+,21-,22+;/m1./s1

InChI key

IZRXCFCWDMSBQY-NXURVHCDSA-N

General description

An antimicrobial agent. A member of the tetracycline family that inhibits bacterial protein synthesis by binding to the 30S ribosomal subunit.
An antimicrobial agent. A member of the tetracycline family. Tetracyclines inhibit bacterial protein synthesis by binding to the 30S ribosomal subunit.

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Other Notes

Hardman, J.G., et al. 1996. in Goodman and Gilman’s The Pharmacological Basis of Therapeutics, Ninth Edition. pp. 1123 - 1125. McGraw-Hill Companies, New York.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Muhammad Abdul Basit et al.
Animals : an open access journal from MDPI, 10(11) (2020-11-25)
The current experiment was designed to estimate the comparative efficacy of selected phytobiotics Persicaria odorata leaf meal (POLM) and Piper betle leaf meal (PBLM) with halquinol, and tetracycline in broiler chickens. The 150-day-old broiler chickens were randomly assigned to five

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service