Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

496260

Sigma-Aldrich

Oncostatin M, Human, Recombinant, E. coli

Synonym(s):

Oncostatin M, Human, Recombinant, E. coli

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.77

recombinant

expressed in E. coli

Quality Level

Assay

≥97% (SDS-PAGE)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤1 EU/μg Endotoxin (EU/μg rhOSM)

shipped in

ambient

storage temp.

−70°C

General description

Recombinant, human oncostatin M expressed in E. coli. Pleiotropic cytokine which inhibits growth of A375 melanoma cells and mouse M1 myeloid leukemic cells. Enhances the growth of both endothelial and hematopoietic cells. Has also been reported to have growth stimulatory activities on normal fibroblasts, AIDS-Kaposis sarcoma cells, and a human erythroleukemic cell line, TF-1.
Recombinant, human oncostatin M expressed in E. coli. Pleiotropic cytokine which inhibits growth of A375 melanoma cells and mouse M1 myeloid leukemic cells. Has also been reported to have growth stimulatory activities on normal fibroblasts, AIDS-Kaposi′s sarcoma cells, and human erythroleukemia cell line, TF-1.

Biochem/physiol Actions

ED₅₀ = 0.15-0.30 ng/ml as measured in a cell proliferation assay using a factor-dependent human erythroleukemic cell line, TF-1

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from sterile-filtered 30% acetonitrile, 0.1% TFA, 50 µg BSA/µg rhOSM.

Reconstitution

Following reconstitution, aliquot and freeze (-70°C). Avoid freeze/thaw cycles of solutions. Stock solutions are stable for up to 3 months at -70°C.
Reconstitute to a final concentration of ≥10 µg/ml in sterile PBS containing ≥0.1% BSA or HSA.

Other Notes

Boileau, C., et al. 2000. J. Immunol.164, 5713.
Miyajima, A., et al. 2000. Cytokine Growth Factor Rev.11, 177.
Linsley, P.S., et al. 1990. Mol. Cell. Biol.10, 1882.
Kitamura, T., et al. 1989. J. Cell Physiol.140, 323.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service