Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

495624

Sigma-Aldrich

Olomoucine

InSolution, ≥98%, potent and selective inhibitor of p34cdc2/cyclin B and related kinases

Synonym(s):

InSolution Olomoucine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H18N6O
Molecular Weight:
298.34
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

shipped in

wet ice

storage temp.

2-8°C

General description

A purine derivative that acts as a potent and selective inhibitor of p34cdc2/cyclin B (IC50 = 7 µM) and related kinases, including p33cdk2/cyclin A (IC50 = 7 µM), p33cdk2/cyclin E (IC50 = 7 µM), p33cdk5/p35 (IC50 = 3 µM), and p44MAPK (IC50 = 25 µM). Acts by competing for the ATP binding domain of the kinase. Exhibits reduced sensitivity towards related kinases, p34cdk4/cyclin D1 (IC50 >1 mM) and p40cdk6/cyclin D3 (IC50 >250 µM). Does not significantly affect the activity of other protein kinases even at 1 mM concentrations. Known to inhibit DNA synthesis in interleukin-2 stimulated T lymphocytes and triggers G1 arrest similar to that observed in interleukin-2 deprived cells. Also used to synchronize cells in G1.

Biochem/physiol Actions

Cell permeable: no
Primary Target
p34cdk1/cyclin B
Product competes with ATP.
Reversible: yes
Target IC50: 100 nM-100 µM against p34cdk1/cyclin B

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 50 mM (5 mg/336 µl) solution of Olomoucine (Cat. No. 495620) in DMSO.

Reconstitution

Following initial use, aliquot and refrigerate (4°C).

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup - (Dimethylsulfoxide)

Flash Point(C)

87 °C - closed cup - (Dimethylsulfoxide)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service