Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

4.91049

Sigma-Aldrich

1-Butyl-3-methylimidazolium tetrafluoroborate

high purity

Synonym(s):

1-Butyl-3-methylimidazolium tetrafluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15BF4N2
CAS Number:
Molecular Weight:
226.02
MDL number:
UNSPSC Code:
12352107

Quality Level

Assay

≥99.0% (HPLC)

pH

5 (20 °C in H2O)

mp

-71 °C

density

1.20 g/cm3

Analysis Note

Purity (HPLC, area%): ≥ 99.0 %
Identity (NMR): passes test
Halides: ≤ 100 ppm
Water (K. F.): ≤ 100 ppm

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service