Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

4.91001

Sigma-Aldrich

1-Ethyl-3-methylimidazolium tris(pentafluoroethyl)trifluorophosphate

high purity

Synonym(s):

1-Ethyl-3-methylimidazolium tris(pentafluoroethyl)trifluorophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11F18N2P
CAS Number:
Molecular Weight:
556.17
UNSPSC Code:
12352107

Quality Level

Assay

≥99.0% (HPLC)

form

liquid

mp

-1 °C

density

1.71 g/cm3 at 20 °C

storage temp.

2-30°C

Analysis Note

Identity (NMR): passes test
Purity (HPLC, area%): ≥ 99.0 %
Halides: ≤ 100 ppm
Water (K. F.): ≤ 100 ppm

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service