Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

489476-M

Millipore

LSD1 Inhibitor

The LSD1 Inhibitor, also referenced under CAS 927019-63-4, controls the biological activity of LSD1. This small molecule/inhibitor is primarily used for Cell Signaling applications.

Synonym(s):

LSD1 Inhibitor, LSD Inhibitor I, BHC110 Inhibitor I, Histone Lysine Demethylase Inhibitor III, KDM1 Inhibitor I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H40Cl4N8
CAS Number:
Molecular Weight:
474.34
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow to yellow

solubility

water: 50 mg/mL

storage temp.

−20°C

General description

A bisguanidine polyamine analogue that exhibits noncompetitive and specific LSD1 inhibition, with 14.1% remaining LSD1 activity at 10 µM in vitro, and is shown to significantly increase methylation at the histone 3 lysine 4 chromatin mark (H3K4me1 and H3K4me2) dose-dependently from 0.25 µM to 10 µM, without affecting global H3K9me2 (which is not a LSD1 substrate) levels in HCT116 human colon carcinoma cells. This compound induces reexpression of several epigenetically silenced genes including SFRP1, SFRP4, SFRP5, and GATA5 more effectively when compared with siRNA treatment, particularly in the case of SFRP4 and SFRP5.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Sharma, SK., et al. 2010 J. Med. Chem.53, 5197.
Huang, Y., et al. 2007. Proc. Natl. Acad. Sci.104, 8023.
Bi, X., et al. 2006. Bioorg. Med. Chem. Lett.16, 3229.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service