Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

487957

Sigma-Aldrich

NOC-18

A nitric oxide (NO) donor similar to NOC-5, NOC-7 and NOC-12 but with an enhanced half-life.

Synonym(s):

NOC-18, DETA/NO, 2,2ʹ-(Hydroxynitrosohydrazino) bis-ethanamine, DETA NONOate, DETA/NO, 2,2ʹ-(Hydroxynitrosohydrazino)bis-ethanamine, DETA NONOate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H13N5O2
CAS Number:
Molecular Weight:
163.18
UNSPSC Code:
12352200

Quality Level

Assay

≥90% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

PBS, pH 7.4: 100 mg/mL
10 mM NaOH: soluble
water: soluble

shipped in

ambient

storage temp.

−20°C

InChI

1S/C4H13N5O2/c5-1-3-8(4-2-6)9(11)7-10/h11H,1-6H2/p+2

InChI key

HMRRJTFDJAVRMR-UHFFFAOYSA-P

General description

A nitric oxide (NO) donor similar to NOC-5 (Cat. No. 487950), NOC-7 (Cat. No. 487952) and NOC-12 (Cat. No. 487955) but with an enhanced half-life. Useful for the controlled release of NO in solution. Induces dose-dependent apoptosis in macrophages. Inhibits lipoprotein oxidation induced by Cu2+in vitro. Half-life (t½) in PBS, pH 7.4 is 3400 minutes at 22°C. Relatively stable in alkaline solution (pH ≥10.0).

Biochem/physiol Actions

Cell permeable: no
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution in NaOH, aliquot and freeze (-20°C). Stock solutions in NaOH are stable for up to 24 h at -20°C. Stock solutions in H₂O or PBS, pH 7.4 are unstable; reconstitute just prior to use.

Other Notes

Seccia, M., et al. 1996. Biochem. Biophys. Res. Commun. 220, 306.
Shibuta, S., et al. 1995. Neurosci. Lett. 187, 103.
Shimaoka, M., et al. 1995. Biochem. Biophys. Res. Commun. 209, 519.
Diodati, J.G., et al. 1993. J. Cardiovasc. Pharmacol. 22, 287.
Hrabie, J.A., et al. 1993. J. Org. Chem. 58,1472.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service