Skip to Content
MilliporeSigma
All Photos(1)

Documents

481408-M

Millipore

NF-κB Activation Inhibitor II, JSH-23

The NF-κB Activation Inhibitor II, JSH-23, also referenced under CAS 749886-87-1, controls the biological activity of NF-κB. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.

Synonym(s):

NF-κB Activation Inhibitor II, JSH-23, 4-Methyl-N¹-(3-phenylpropyl)benzene-1,2-diamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H20N2
CAS Number:
Molecular Weight:
240.34
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 10 mg/mL

storage temp.

−20°C

InChI

1S/C16H20N2/c1-13-9-10-16(15(17)12-13)18-11-5-8-14-6-3-2-4-7-14/h2-4,6-7,9-10,12,18H,5,8,11,17H2,1H3

InChI key

YMFNPBSZFWXMAD-UHFFFAOYSA-N

General description

A cell-permeable diamino compound that selectively blocks nuclear translocation of NF-κB p65 and its transcription activity (IC50 = 7.1 µM in a NF-κB reporter assay using RAW 264.7) without affecting IκB degradation. Shown to suppress DNA-binding of NF-κB and downregulate LPS-induced gene expression and apoptotic chromatin condensation.
A cell-permeable, selective blocker of the nuclear translocation of NF-κB p65 that does not affect IκB degradation (IC50 = 7.1 µM in LPS-stimulated macrophages RAW 264.7 stably transfected with pNF-κB-SEAP-NPT). Shown to suppress DNA-binding of NF-κB and downregulate LPS-inducible expression of TNF-α, IL-1β, IL-6, iNOS, and COX-2. Also prevents LPS-induced apoptotic chromatin condensation.

Biochem/physiol Actions

Primary Target
Blocks nuclear translocation of NF-κB p65 and its transcription activity
Target IC50: 7.1 µM in a NF-κB reporter assay using RAW 264.7

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Shin, H.M., et al. 2004. FEBS Lett.571, 50.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service