Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

480748

Sigma-Aldrich

Neuronal Differentiation Inducer, Isobavachin

A cell-permeable, 8-prenyl flavone that is shown to induce differentiation of mouse embryonic stem (ES) cells into neuronal cells and astrocytes, at 100 nM.

Synonym(s):

Neuronal Differentiation Inducer, Isobavachin, 7-hydroxy-2-(4-hydroxyphenyl)-8-(3-methylbut-2-enyl)chroman-4-one, Isobavachin, Isobavachin, 7-hydroxy-2-(4-hydroxyphenyl)-8-(3-methylbut-2-enyl)chroman-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H20O4
CAS Number:
Molecular Weight:
324.37
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow

solubility

DMSO: 100 mg/mL, clear, yellow

shipped in

ambient

storage temp.

−20°C

SMILES string

OC1=CC=C(C(CC(C2=CC=C(O)C=C2)O3)=O)C3=C1CC=C(C)C

General description

A cell-permeable, 8-prenyl flavone that is shown to induce differentiation of mouse embryonic stem (ES) cells into neuronal cells and astrocytes, at 100 nM. It is shown to elicit the down-regulation of p38 and JNK phosphorylation, and the up-regulation of ERK phosphorylation. Co-treatment with GGTI-298 (1000 nM), a Geranylgeranyltransferase I Inhibitor (Cat. No. 345883) selectively abolishes the IBA- but not Retinoic Acid-induced neuronal differentiation, which suggests that the differentiation inducing effect may involve protein prenylation.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Wang, D.Y., et al. 2011. Acta Pharmacol Sin32, 425.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service