Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

480746-M

Millipore

Neuronal Differentiation Inducer IV

A cell-permeable quinoxaline compound that is shown to induce neuronal differentiation in PC12 and rat MSC (mesenchymal stem cell) cultures with high efficiency.

Synonym(s):

Neuronal Differentiation Inducer IV, N-Hydroxy-2-oxo-3-(3-phenylpropyl)-1,2-dihydroquinoxaline-6-carboxamide, 2-Oxo-3-(3-phenyl-propyl)-1,2-dihydro-quinoxaline-6-carboxylic acid hydroxyamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H17N3O3
Molecular Weight:
323.35
UNSPSC Code:
12352200

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 25 mg/mL, pale yellow

storage temp.

2-8°C

General description

A cell-permeable quinoxaline compound that is shown to induce neuronal differentiation in PC12 and rat MSC (mesenchymal stem cell) cultures with high efficiency (>95%; 20 µM for 48 h) as determined by Nissl staining, expressions of neuronal markers and genes (β-III tubulin, NSE, Chrna2, Chrnb2, Chrm4, Foxo2, Sizn 1), as well as by whole-cell electrophysiology measurements, without any concomitant astroglial differentiation induction as indicated by the absence of GFAP (glial fibrillary acidic protein) expression.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Kim, N.R., et al. 2009. J. Med. Chem.52, 7931.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service