4.80680
Hydrochloric acid 3 Mol/L
EMPROVE® EXPERT
Pharma Manufacturing
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
Quality Level
product line
EMPROVE® EXPERT
pH
<1 (20 °C in H2O)
density
1.05 g/cm3 at 20 °C
storage temp.
2-25°C
Related Categories
General description
EMPROVE® EXPERT
Legal Information
Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
also commonly purchased with this product
related product
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
8B - Non-combustible, corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service