Skip to Content
MilliporeSigma
All Photos(1)

Documents

480420

Sigma-Aldrich

NF449

A potent Gsα-subunit-selective G-protein antagonist.

Synonym(s):

NF449, 4,4ʹ,4ʹʹ,4ʹʹʹ-[Carbonyl- bis[imino-5,1,3-benzenetriyl bis-(carbonylimino)]]tetrakis-(benzene-1,3-disulfonic Acid, 8Na)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H24N6O29S8 · 8Na
CAS Number:
Molecular Weight:
1505.09
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

distilled water: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C41H32N6O29S8.8Na/c48-37(44-29-5-1-25(77(53,54)55)15-33(29)81(65,66)67)19-9-20(38(49)45-30-6-2-26(78(56,57)58)16-34(30)82(68,69)70)12-23(11-19)42-41(52)43-24-13-21(39(50)46-31-7-3-27(79(59,60)61)17-35(31)83(71,72)73)10-22(14-24)40(51)47-32-8-4-28(80(62,63)64)18-36(32)84(74,75)76;;;;;;;;/h1-18H,(H,44,48)(H,45,49)(H,46,50)(H,47,51)(H2,42,43,52)(H,53,54,55)(H,56,57,58)(H,59,60,61)(H,62,63,64)(H,65,66,67)(H,68,69,70)(H,71,72,73)(H,74,75,76);;;;;;;;/q;8*+1/p-8

InChI key

KCBZSNWCUJBMHF-UHFFFAOYSA-F

General description

A potent G-selective G-protein antagonist. Suppresses the rate of GTPγS binding to Gsα-s (IC50 = 140 nM) but not in Giα-1. Inhibits stimulation of adenylate cyclase activity in S49 cyc cells (which lack endogenous G) by exogenously added Gsα-s. Also blocks the coupling of β-adrenergic receptors to Gs (EC50 = 7.9 µM). Acts as a reversible competitive antagonist of the human P2X receptor, exhibiting much higher potency at human P2X(1) than at P2X(7) receptors (IC50 = 50 nM vs 40 µM respectively).
A potent Gsα-subunit-selective G-protein antagonist. Suppresses the rate of GTPγS binding to Gsα-s-subunit (IC50 = 140 nM) but not to Giα-1-subunit. Inhibits stimulation of adenylate cyclase activity in S49 cyc- cells (which lack endogenous Gsα-subunit) by exogenously added Gsα-subunit. Also blocks the coupling of β-adrenergic receptors to Gs (EC50 = 7.9 µM).

Biochem/physiol Actions

Cell permeable: no
Primary Target
Gsα-subunit-selective G-protein antagonist
Product does not compete with ATP.
Reversible: no
Target IC50: 140 nM suppressing the rate of GTPγS binding to Gsα-s-subunit

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Hulsmann, M., et al. 2003. Eur. J. Pharmacol.470, 1.
Hohenegger, M., et al. 1998. Proc. Natl. Acad. Sci. USA 95, 346.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaozhe Zhang et al.
The Journal of biological chemistry, 295(30), 10281-10292 (2020-06-05)
Inositol hexakisphosphate (IP6) is an abundant metabolite synthesized from inositol 1,3,4,5,6-pentakisphosphate (IP5) by the single IP5 2-kinase (IP5K). Genetic and biochemical studies have shown that IP6 usually functions as a structural cofactor in protein(s) mediating mRNA export, DNA repair, necroptosis
Kazue Hisaoka-Nakashima et al.
Journal of neurochemistry, 158(4), 849-864 (2020-10-30)
Lysophosphatidic acid (LPA), a brain membrane-derived lipid mediator, plays important roles including neural development, function, and behavior. In the present study, the effects of LPA on astrocyte-derived synaptogenesis factor thrombospondins (TSPs) production were examined by real-time PCR and western blotting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service