Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

480275

Sigma-Aldrich

β-NGF, Human, Recombinant, NSO Cells

Synonym(s):

β-Nerve Growth Factor, Human, Recombinant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.75

recombinant

expressed in NSO cells

Quality Level

Assay

≥97% (SDS-PAGE)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤100 pg/μg Endotoxin (pg/μg β-NGF)

shipped in

ambient

storage temp.

−70°C

General description

Recombinant, human β-nerve growth factor expressed in NSO cells. Involved in neuroimmune interactions and inflammation.

Biochem/physiol Actions

ED₅₀ = 800-1500 pg/ml as measured in a cell proliferation assay using a factor-dependent human erythroleukemic cell line

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from sterile-filtered solution in PBS containing 50 µg BSA/µg β-NGF.

Reconstitution

Following reconstitution, aliquot and freeze (-70°C). Avoid freeze/thaw cycles of solutions. Stock solutions are stable for up to 3 months at -70°C.
Reconstitute to a concentration ≥1 µg/ml with sterile PBS containing at least 0.1% HSA or BSA.

Other Notes

Leon, A., et al. 1994. Proc. Natl. Acad. Sci. USA91, 3739.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service