Skip to Content
MilliporeSigma
All Photos(1)

Documents

480060

Sigma-Aldrich

Necrosis Inhibitor, IM-54

The Necrosis Inhibitor, IM-54, also referenced under CAS 861891-50-1, selectively blocks oxidative stress-induced necrotic cell death. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

Necrosis Inhibitor, IM-54, 2-(1H-Indol-3-yl)-3-pentylamino-maleimide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H23N3O2
CAS Number:
Molecular Weight:
325.40
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow-orange

solubility

methanol: 10 mg/mL
DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C19H23N3O2/c1-4-5-8-11-20-17-16(18(23)22(3)19(17)24)14-12-21(2)15-10-7-6-9-13(14)15/h6-7,9-10,12,20H,4-5,8,11H2,1-3H3

InChI key

SGLOMINNEBLJFF-UHFFFAOYSA-N

General description

A cell-permeable mono-indolylmaleimide compound that selectively blocks oxidative stress-induced necrotic cell death (~3 µM IM-54 prevented ~50% cell death in HL60 cells exposed to 100 µM H2O2). Does not offer protection against Etoposide- (Cat. No. 341205) induced apoptosis or display antioxidant properties. Does not affect the kinase activities of S6K1 and PKC isozymes even at concentrations as high as 50 µM.
A cell-permeable mono-indolylmaleimide compound that selectively blocks oxidative stress-induced necrotic cell death (~3 µM IM-54 prevented ~50% cell death in HL60 cells exposed to 100 µM H2O2). Does not offer protection against Etoposide- (Cat. No. 341205) induced apoptosis or display antioxidant properties. Does not affect the kinase activities of S6K1 and PKC isozymes even at concentrations as high as 50 µM.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Blocks oxidative stress-induced necrotic cell death
Product does not compete with ATP.
Reversible: no
Target IC50: (~3 µM prevented ~50% cell death in HL60 cells exposed to 100 µM H₂O₂).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Dodo, K., et al. 2005. Bioorg. Med. Chem. Lett.15, 3114.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toria Tomasek et al.
Biochemical and biophysical research communications, 556, 199-206 (2021-04-14)
Circulating cell-free hemoglobin (CFH) contributes to endothelial injury in several inflammatory and hemolytic conditions. We and others have shown that CFH causes increased endothelial permeability, but the precise mechanisms of CFH-mediated endothelial barrier dysfunction are not fully understood. Based on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service