Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

475914

Sigma-Aldrich

Nystatin

from Streptomyces noursei, ≥95% (sum of two peaks, HPLC), solid, sodium ionoph, Calbiochem®

Synonym(s):

Nystatin, Streptomyces noursei, Mycostatin

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$85.50
5 G
$259.00

$85.50


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$85.50
5 G
$259.00

About This Item

Empirical Formula (Hill Notation):
C47H75NO17
CAS Number:
Molecular Weight:
926.09
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$85.50


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

Nystatin, Streptomyces noursei, A polyene antifungal antibiotic effective against yeast and mycoplasma.

Quality Level

description

Merck USA index - 14, 6736

Assay

≥95% (sum of two peaks, HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white to yellow

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

N[C@@H]1[C@@H]([C@H](O[C@@H]([C@H]1O)C)OC2CC3O[C@@](CC(C3C(=O)O)O)(CC(C(CCC(CC(CC(CC(=O)O[C@H]([C@@H]([C@@H]([C@H](\C=C\C=C\CC\C=C\C=C\C=C\C=C\2)C)O)C)C)O)O)O)O)O)O)O

InChI

1S/C47H75NO17/c1-27-17-15-13-11-9-7-5-6-8-10-12-14-16-18-34(64-46-44(58)41(48)43(57)30(4)63-46)24-38-40(45(59)60)37(54)26-47(61,65-38)25-36(53)35(52)20-19-31(49)21-32(50)22-33(51)23-39(55)62-29(3)28(2)42(27)56/h5-6,8,10-18,27-38,40-44,46,49-54,56-58,61H,7,9,19-26,48H2,1-4H3,(H,59,60)/b6-5+,10-8+,13-11+,14-12+,17-15+,18-16+/t27-,28-,29-,30+,31?,32?,33?,34?,35?,36?,37?,38?,40?,41-,42+,43+,44-,46+,47+/m0/s1

InChI key

VQOXZBDYSJBXMA-RKEBNKJGSA-N

General description

A polyene antifungal antibiotic effective against yeast and mycoplasma. Induces loss of low molecular weight substances from cells with low selectivity. A cation selective ionophore that exhibits selectivity for Na+. Increases the activity of the Na+-K+ pump. Suggested concentration is 50 µg/ml.

Biochem/physiol Actions

Potency: ≥3000 units/mg (anhydrous).

Warning

Toxicity: Standard Handling (A)

Other Notes

Gruwel, M.L., et al. 1995. Am J. Physiol.268, H351.
Opekarova, M., and Tanner, W. 1994. FEBS Lett. 360, 46.
Stoos, B.A., et al. 1994. J. Am. Soc. Nephrol.4, 1855.
Skolink, P.R., et al. 1989. J. Infect. Dis.160, 1056.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service