Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

475899

Millipore

MOPS sodium salt

ULTROL® Grade, ≥98% dry basis (titration), zwitterionic buffer useful in pH range 6.5-7.9

Synonym(s):

MOPS, Sodium, ULTROL® Grade, 3-(N-Morpholino)propanesulfonic Acid, Na

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14NO4S · Na
CAS Number:
Molecular Weight:
231.25
MDL number:
UNSPSC Code:
12161700
NACRES:
NA.21
Pricing and availability is not currently available.

description

Merck USA index - 14, 6265

Quality Level

Assay

≥98% dry basis (titration)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 1 M (232 mg/ml)

cation traces

heavy metals: ≤1 ppm

shipped in

ambient

storage temp.

15-25°C

SMILES string

[Na+].[S](=O)(=O)([O-])CCCN1CCOCC1

InChI

1S/C7H15NO4S.Na/c9-13(10,11)7-1-2-8-3-5-12-6-4-8;/h1-7H2,(H,9,10,11);/q;+1/p-1

InChI key

MWEMXEWFLIDTSJ-UHFFFAOYSA-M

General description

A zwitterionic buffer useful in the pH range of 6.5-7.9. Has a pKa of 7.20 at 25°C.
Absorbance (0.1 M, H2O, 260 nm and 280 nm):≤0.040.
Zwitterionic buffer active in the pH range of 6.5-8.0. Absorbance: ≤0.04 at 260 nm, 1.0 M, H2O; pKa=7.2 at 20°C.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, store at room temperature (15-33°C). Stock solutions are stable for up to 6 months at room temperature.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTROL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service