Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

475897

Sigma-Aldrich

Monensin Methyl Ester

A neutral analog of monensin.

Synonym(s):

Monensin Methyl Ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H64O11
Molecular Weight:
684.90
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

color

white

solubility

ethanol: 68 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A neutral analog of monensin. Useful in the preparation of ion-selective membranes for ion-selective electrodes.
For use in the preparation of ion-selective membranes for ion-selective electrodes.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Useful in the preparation of ion-selective membranes for ion-selective electrodes
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Highly Toxic (H)

Reconstitution

Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for up to 6 months at 4°C. Unstable in acidic solutions.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Schindler, J.G., and Schindler, M.M. 1987. Biomed. Tech. (Berl.)32, 22.
Chamberlin, J.W. 1978. Org. Mass Spect.3, 271.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service