Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

475859

Sigma-Aldrich

Mitochondrial Fusion Promoter, M1

A cell-permeable phenylhydrazone that restores mitochondrial tubular network formation in MEF lacking either of the two outer mitochondrial membrane.

Synonym(s):

Mitochondrial Fusion Promoter, M1, ( E)-4-Chloro-2-(1-(2-(2,4,6-trichlorophenyl)hydrazono)ethyl)phenol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10Cl4N2O
CAS Number:
Molecular Weight:
364.05
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 25 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

ClC1=CC(/C(C)=N/NC2=C(Cl)C=C(Cl)C=C2Cl)=C(O)C=C1

General description

A cell-permeable phenylhydrazone compound that restores mitochondrial tubular network formation from the fragmented mitochondria seen in MEF lacking either one of the two outer mitochondrial membrane (OMM) mitofusins (EC50 = 5.3 and 4.42 µM, respectively, in Mfn1 or Mfn2 knockout MEF cells) or MPP+-treated SH-SY5Y cells (5 µM 24 h), while displaying no effect on ER or lysosome morphology in Mfn1 knockout MEF. The effect of M1 is limited to enhancing weakened mitochondrial fusion machinery and M1 cannot by itself rebuild interconnected tubular mitochondria in MEF lacking both Mfn1/2 or the inner mitochondrial membrane (IMM) fusion mediator Opa1 (optic atrophy1). M1 (5 µM 24 h) is reported to boost the downregulated ATP5A and ATP5B protein level in either Mfn1 or Mfn2 knockout MEF to the wild-type MEF level and ATPase inhibitor oligomycin (Cat. No. 495455) at 5 µM is shown to completely offset the mitochondrial fusion effect by 5 µM M1 in Mfn1 knockout MEF. Both M1 and Z-VAD-FMK (Cat. No. 219007) are shown to protect SH-SY5Y against MPP+-induced neuronal toxicity and additive protection can be achieved via a combined treatment (62%, 73%, 77%, and 89% survival rate, respectively, with DMSO, 5 µM M1, 1 µM Z-VAD-FMK, and combined treatment). Comparing to mdivi-1 (Cat. No. 475856), M1 exerts its effect via promoting fusion rather than inhibiting fission or division.
A cell-permeable phenylhydrazone that restores mitochondrial tubular network formation in MEF lacking either of the two outer mitochondrial membrane (OMM) mitofusins (EC50 = 5.3 and 4.42 µM, respectively, in Mfn1 or Mfn2 knockout MEF cells) or in MPP+-treated SH-SY5Y cells (5 µM 24 h), but not in MEF lacking both Mfn1/2 or the inner mitochondrial membrane (IMM) fusion mediator Opa1 (optic atrophy1). M1 (5 µM 24 h) is reported to boost the downregulated ATP5A & ATP5B protein level in Mfn1 or Mfn2 knockout MEF to the wild-type MEF level and ATPase inhibitor oligomycin (Cat. No. 495455) at 5 µM is shown to completely offset the mitochondrial fusion effect by 5 µM M1 in Mfn1 knockout MEF. Comparing to mdivi-1 (Cat. No. 475856), M1 exerts its effect via promoting fusion rather than inhibiting fission or division.

Biochem/physiol Actions

Cell permeable: yes
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Wang, D., et al. 2012. Angew. Chem. Int. Ed.51, 9302.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A small molecule promotes mitochondrial fusion in mammalian cells.
Danling Wang et al.
Angewandte Chemie (International ed. in English), 51(37), 9302-9305 (2012-08-22)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service