Skip to Content
MilliporeSigma
All Photos(1)

Documents

474427

Sigma-Aldrich

4-Methylumbelliferyl-β-D-glucuronide - CAS 6160-80-1 - Calbiochem

Substrate for fluorogenic assay of β-D-glucuronidase.

Synonym(s):

4-Methylumbelliferyl-β-D-glucuronide - CAS 6160-80-1 - Calbiochem, MUG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16O9
CAS Number:
Molecular Weight:
352.29
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥99% (TLC)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

impurities

<25 ppm 4-methylumbelliferone

color

white

solubility

aqueous buffer: soluble
methanol: soluble
water: soluble

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C16H16O9/c1-6-4-10(17)24-9-5-7(2-3-8(6)9)23-16-13(20)11(18)12(19)14(25-16)15(21)22/h2-5,11-14,16,18-20H,1H3,(H,21,22)/t11-,12-,13+,14-,16+/m0/s1

InChI key

ARQXEQLMMNGFDU-JHZZJYKESA-N

General description

This product has been discontinued.



Substrate for fluorogenic assay of β-D-glucuronidase.

Substrate for fluorometric assay of β-D-glucuronidase (excitation maximum: 360 nm; emission maximum: 440 nm).

Warning

Toxicity: Standard Handling (A)

Other Notes

Moberg, L.J. 1985. Appl. Environ. Microbiol. 50, 1383.
Trepeta, R.W., and Edberg, S.C. 1984. J. Clin. Microbiol. 19, 172.
Brot, F.E., et al. 1974. Biochem. Biophys. Res. Commun. 57, 1.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service