Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

454862

Sigma-Aldrich

MNK1/2 Inhibitor II, ETP-45835

The MNK1/2 Inhibitor II, ETP-45835 controls the biological activity of MNK1/2. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

MNK1/2 Inhibitor II, ETP-45835, 4-(3-(Piperidin-4-yl)-1H-pyrazol-5-yl)pyridine, diHCl, ETP45835, MNK Inhibitor II, Mitogen-Activated Protein Kinase-Interacting Kinase Inhibitor II, ETP45835, MNK Inhibitor II, Mitogen-Activated Protein Kinase-Interacting Kinase Inhibitor II, 4-(3-(Piperidin-4-yl)-1H-pyrazol-5-yl)pyridine, diHCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16N4 · 2HCl
CAS Number:
Molecular Weight:
301.21
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

water: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

C1(C2=CC=NC=C2)=CC(C3CCNCC3)=NN1

General description

A cell-permeable 3,5-disubstituted pyrazolo compound that acts as a selective MNK inhibitor (IC50 = 646 and 575 nM, respectively, against MNK1 and MNK2) with much reduced or little activity against a panel of 24 other kinases (≤15% inhibition at 5 µM), including MNK upstream kinases B-Raf, ERK1, MEK1, and p38α. Reported to exhibit good aqueous solubility (>100 µM at pH 7.4) and inhibit the proliferation (IC50 = 17 µM) and cellular eIF4E Ser209 phosphorylation (IC50 = 4.7 µM) of acute myeloid leukemia MV4:11 cells.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Oyarzabal, J., et al. 2010. J. Med. Chem.53, 6618.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service