Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

454583-M

Sigma-Aldrich

PhosphoDetect Anti-NMDA NR2B (pTyr¹⁴⁷²) Rabbit pAb

liquid, Calbiochem®

Synonym(s):

Anti-N-Methyl-D-Aspartate Receptor Subunit 2B, Phospho-Specific (Tyr¹⁴⁷²)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

does not contain

preservative

species reactivity

rat

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr1472)

Gene Information

General description

Immunoaffinity purified rabbit polyclonal antibody. Recognizes the ~180 kDa NR2B subunit of the NMDA receptor phosphorylated at Tyr1472.
Recognizes the ~180 kDa NMDA receptor NR2B subunit phosphorylated at Tyr1472.

Immunogen

Rat
a synthetic phosphopeptide corresponding to amino acids surrounding the Tyr¹⁴⁷² phosphorylation site of rat NMDA NR2B

Application

Dot Blot (1:500-1:1000)

Immunoblotting (1:500-1:1000)

Warning

Toxicity: Standard Handling (A)

Physical form

In 150 mM NaCl, 10 mM HEPES, 100 µg/ml BSA, 50% glycerol, pH 7.5.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

May detect 2 additional lower molecular weight bands. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service