Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

4510-OP

Sigma-Aldrich

OmniPur® Fast Green FCF

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$211.00

$211.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 G
$211.00

About This Item

UNSPSC Code:
12171500
NACRES:
NA.28

$211.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Legal Information

OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Muta. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

352.4 °F - closed cup

Flash Point(C)

178 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service