Skip to Content
MilliporeSigma
All Photos(1)

Documents

444905

Sigma-Aldrich

Anti-Mast Cell Tryptase Mouse mAb (AA1)

liquid, clone AA1, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

AA1, monoclonal

form

liquid

does not contain

preservative

species reactivity

monkey, canine, human, feline

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TPSAB1(7177)

General description

Protein G purified mouse monoclonal antibody. Recognizes the ~32-38 kDa α- and β-isoforms of tryptase in mast cells.
Recognizes the 32-38 kDa α- and β-isoforms of tryptase in mast cells.
This Anti-Mast Cell Tryptase Mouse mAb (AA1) is validated for use in Frozen Sections, Immunoblotting, Paraffin Sections for the detection of Mast Cell Tryptase.

Immunogen

Human
native, human mast cell, tryptase

Application

Frozen Sections (0.06-0.12 µg/ml)

Immunoblotting (1 µg/ml)

Paraffin Sections (0.06-0.12 µg/ml, heat pre-treatment required, see comments)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 10 mM PBS, pH 7.4.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
Cat, dog, human, or monkey mast cells

Other Notes

Sommerhoff, C.P., et al. 2000. Biochim. Biophys. Acta.1477, 75.
Little, S.S., and Johnson, D.A. 1995. Biochem. J.307, 341.
Morgan, S.J., et al. 1991. J. Allergy Clin. Immunol.87, 111.
Walls, A.F., et al. 1990. J. Pathol.162, 119.
Wall, A.F., et al. 1990. Clin. Exp. Allergy20, 581.
Staining of formalin-fixed paraffin sections requires boiling tissue sections in 10 mM citrate buffer, pH 6.0, for 10-20 min followed by cooling at room temperature for 20 min. Variables associated with assay conditions will dictate the optimal working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Johannes Kapeller et al.
The Journal of comparative neurology, 519(3), 420-432 (2010-12-31)
Since the first description of 5-HT₃ receptors more than 50 years ago, there has been speculation about the molecular basis of their receptor heterogeneity. We have cloned the genes encoding novel 5-HT3 subunits 5-HT3C, 5-HT3D, and 5-HT3E and have shown
Henry R Maun et al.
Cell, 179(2), 417-431 (2019-10-05)
Severe asthma patients with low type 2 inflammation derive less clinical benefit from therapies targeting type 2 cytokines and represent an unmet need. We show that mast cell tryptase is elevated in severe asthma patients independent of type 2 biomarker

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service