Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

444248

Sigma-Aldrich

MMP-13, Proenzyme, His•Tag®, Human, Recombinant, S. frugiperda

Synonym(s):

Collagenase-3

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$947.00

$947.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
$947.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

$947.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥90% (SDS-PAGE)

Quality Level

form

liquid

specific activity

≥50 mU/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

General description

Full-length recombinant, human MMP-13 fused to a His•Tag sequence and expressed in S. frugiperda insect cells. This 452 amino acid proenzyme contains an N-terminal propeptide which confers latency to the proenzyme, a Ca2+- and Zn2+- binding catalytic domain, a hinge region, and a C-terminal hemopexin domain. Hydrolyzes collagen type II 5-6 times faster than collagens type I and III. Exhibits high activity towards gelatin and degrades α1-antichymotrypsin and plasminogen activator inhibitor-2.
Note: 1 mU = 1 milliunit.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of APMA-activated enzyme that will hydrolyze 1.0 µmol MCA-Pro-Leu-Gly-Leu-Dpa-Ala-Arg-NH₂ (Cat. No. 03-32-5032) per min at 37°C, pH 7.5.

Physical form

In 150 mM NaCl, 50 mM Tris-HCl, 5 mM CaCl₂, pH 7.5.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Herget, J., et al. 2003. Am. J. Physiol.285, 4199.
Nagase, H. 1997. Biol. Chem.378, 151.
Knäuper, V., et al. 1996. J. Biol. Chem. 271, 1544.
Knauper, V., et al. 1996. J. Biol. Chem.271, 17124.
Pei, D., and Weiss, S.J. 1996. J. Biol. Chem.271, 9135.
Knight, C.G., et al. 1992. FEBS Lett.296, 263.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
HIS TAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service