Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

442685-M

Millipore

PhosphoDetect Anti-MAP Kinase ERK1/ERK2 (pThr²⁰²/Tyr²⁰⁴) Rabbit pAb

liquid, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

does not contain

preservative

species reactivity

human, rat

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

storage temp.

−20°C

target post-translational modification

phosphorylation (pThr202/pTyr204)

Gene Information

human ... MAPK3(5595)
rat ... Mapk3(50689)

General description

Immunoaffinity purified rabbit polyclonal antibody. Recognizes the ~42-44 kDa ERK1 and ERK2 proteins phosphorylated at Thr202 and Tyr204.
Recognizes the ~42-44 kDa ERK1 and ERK2 proteins phosphorylated at Thr202 and Tyr204.
This PhosphoDetect Anti-MAP Kinase ERK1/ERK2 Rabbit pAb is validated for use in Dot Blot, WB for the detection of MAP Kinase ERK1/ERK2 .

Immunogen

a synthetic phosphopeptide surrounding the Thr²⁰²/Tyr²⁰⁴ phosphorylation sites of human ERK1/ERK2

Application

Dot Blot (1:1000)

Immunoblotting (1:1000)

Warning

Toxicity: Standard Handling (A)

Physical form

In 150 mM NaCl, 10 mM HEPES, 100 mg/ml BSA, 50% glycerol, pH 7.5.

Other Notes

Does not react with the unphosphorylated proteins. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service