Skip to Content
MilliporeSigma
All Photos(1)

Documents

440319

Sigma-Aldrich

Lumican, His•Tag®, CF, Mouse, Recombinant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.45

Assay

≥95% (SDS-PAGE)

Quality Level

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤1.0 EU/μg protein Endotoxin

shipped in

ambient

storage temp.

−20°C

General description

Recombinant, murine lumican, consisting of amino acids 19-338, fused at the C-terminus to a His•Tag sequence, and expressed in NS0-derived mouse myeloma cells. Lumincan a 40-kDa member of the small leucine-rich repeat proteoglycans (SLRPs) family/Class II subfamily of proteins. It is known for its ability to modulate collagen fibrillogenesis and role in cell proliferation regulation.

Biochem/physiol Actions

Cell adhesion: 50-70% after 50 min at 37°C as measured by the ability of immobilized lumican to supoprt the adhesion of 5 X 10⁴ BCE C/D-1b bovine cornea-derived endothelial cells.

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 0.2 µm filtered PBS.

Reconstitution

Following reconstitution refrigerate (4°C) for short-term storage or aliquot and freeze (-20°C) for long-term storage. Stock solutions are stable for up to 1 month at 4°C or for up to 3 months at -20°C.
Reconstitute to 200 µg/ml with sterile PBS.

Other Notes

D′Onofrio, M.F. et al. 2008. Biochem. Biophys. Res. Commun.365, 266.
Nikitovic, D. et al. 2008. IUBMB Life60, 818.
Nikitovic, D. et al. 2008. FEBS J.275, 350.
Ishiwata, T. et al. 2007. Oncol. Rep.18, 537.
Vuillermoz, B. et al. 2004. Exp. Cell Res.296,294.
Chakravarti, S. et al. 2003. Invest. Ophthalmol. Vis. Sci.44, 2422.
Jepsen, K.J. et al. 2002. J. Biol. Chem.277, 35532.
Chakravarti, S. et al. 2000. Invest. Ophthalmol. Vis. Sci.41,3365.
Blochberger, T.C. et al. 1992. J. Biol. Chem.267, 347.
Chakravarti, S. et al. 1998. J. Cell Biol.141, 1277.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
HIS TAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service