Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

438210

Sigma-Aldrich

LSF Inhibitor, FQI1

The LSF Inhibitor, FQI1 controls the biological activity of LSF.

Synonym(s):

LSF Inhibitor, FQI1, (±)(8-(2-Ethoxyphenyl)-7,8-dihydro-[1,3]dioxolo[4,5-g]quinolin-6(5H)-one, Factor Quinolinone Inhibitor 1, Racemic

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H17NO4
Molecular Weight:
311.33
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.28

Quality Level

Assay

>92% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

A cell-permeable dihydroquinolinone compound that acts as a potent, specific, and reversible inhibitor of DNA-binding activity of Late SV40 Factor (LSF) and blocks LSF transactivation (IC50 = 2.1 µM in NIH 3T3 cells). Exhibits anti-proliferative activity and growth arrest in multiple cell lines (GI50 = 3.8, 0.79, and 6.3 µM in NIH 3T3, HeLa S3 and A549 cells, respectively). Shown to induce apoptotic cell death in liver cancer cell lines over-expressing LSF without affecting primary or immortalized hepatocytes. Also shown to inhibit hepatocellular carcinoma s.c. growth in mouse xenograft model without any toxic effects (1 mg/kg, i.p., once every 3 days).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Grant, T.J., et al. 2012. Proc. Natl. Acad. Sci. USA109, 4503.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service