Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

436305-M

Millipore

Linoleic Acid

An essential fatty acid.

Synonym(s):

Linoleic Acid, cis,cis-9,12-Octadecadienoic Acid, Linolic Acid, Linolic Acid, cis,cis-9,12-Octadecadienoic Acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H32O2
CAS Number:
Molecular Weight:
280.45
MDL number:
UNSPSC Code:
12352211

Assay

≥97% (LC/MS)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

colorless

solubility

ethanol: 1 mg/mL

storage temp.

2-8°C

InChI

1S/C18H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h6-7,9-10H,2-5,8,11-17H2,1H3,(H,19,20)/b7-6-,10-9-

InChI key

OYHQOLUKZRVURQ-HZJYTTRNSA-N

General description

An essential fatty acid.
An essential fatty acid. Density: 900 mg/ml.
Density: 900 mg/ml.

Biochem/physiol Actions

Primary Target
Essential fatty acid

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service