Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

434625

Sigma-Aldrich

Leukotriene B₄

≥97% (HPLC), oil, proinflammatory agent, Calbiochem®

Synonym(s):

Leukotriene B4, LTB₄, 5(S),12(R)-Dihydroxy-6- cis-8- trans-10- trans-14- cis-eicosatetraenoic Acid

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$609.00

$609.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 μG
$609.00

About This Item

Empirical Formula (Hill Notation):
C20H32O4
CAS Number:
Molecular Weight:
336.47
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.77

$609.00


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

Leukotriene B4, Potent proinflammatory agent, primarily because of its actions on polymorphonuclear leukocytes that include chemotaxis, aggregation, degranulation, and stimulation of adherence to vascular endothelium.

description

Merck USA index - 14, 5455

Quality Level

Assay

≥97% (HPLC)

form

oil

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

clear

solubility

ethanol: soluble

shipped in

wet ice

storage temp.

−70°C

SMILES string

OC(CCCC(=O)O)\C=C\C=C\C=C\C(O)C\C=C\CCCCC

InChI

1S/C20H32O4/c1-2-3-4-5-6-9-13-18(21)14-10-7-8-11-15-19(22)16-12-17-20(23)24/h6-11,14-15,18-19,21-22H,2-5,12-13,16-17H2,1H3,(H,23,24)/b8-7+,9-6+,14-10+,15-11+

InChI key

VNYSSYRCGWBHLG-XUOUMLBJSA-N

General description

Potent inflammatory agent, primarily because of its actions on polymorphonuclear leukocytes that include chemotaxis, aggregation, degranulation, and stimulation of adherence to vascular endothelium. Stimulates c-fos and c-jun proto-oncogene transcription in human monocytes. Involved in the pathogenesis of postischemic cerebral edema.
Potent proinflammatory agent, primarily because of its actions on polymorphonuclear leukocytes that include chemotaxis, aggregation, degranulation, and stimulation of adherence to vascular endothelium. Stimulates c-fos and c-jun proto-oncogene transcription in human monocytes. Involved in the pathogenesis of postischemic cerebral edema and asthma.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Polymorphonuclear leukocytes
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Flammable (J)

Physical form

In Ethanol.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Kendall, A.P., et al. 1995. Anesthesia 50, 590.
Dokter, W.H., et al. 1994. Leukemia 8, 1181.
Jedlitschky, G., et al. 1991. J. Biol. Chem.266, 24763.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service