Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

428019-M

Millipore

Anti-Lamp2 Rat mAb (GL2A7)

liquid, clone GL2A7, Calbiochem®

Synonym(s):

Anti-Lysosome-Associated Membrane Protein 2, Anti-Igp-110

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rat

antibody form

ascites fluid

antibody product type

primary antibodies

clone

GL2A7, monoclonal

form

liquid

does not contain

preservative

species reactivity

mouse

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Lamp2(16784)

General description

Rat polyclonal antibody supplied as undiluted ascites. Recognizes the ~60 kDa Lamp2 protein.
Recognizes the ~60 kDa Lamp2 protein.
This Anti-Lamp2 Rat mAb (GL2A7) is validated for use in Immunoblotting, Immunocytochemistry, Immunoprecipitation for the detection of Lamp2.

Immunogen

Mouse
mouse liver lysosomal membranes

Application

Immunoblotting (1:200)

Immunocytochemistry (1:100, see application references)

Immunoprecipitation (see application references)

Warning

Toxicity: Standard Handling (A)

Physical form

Undiluted ascites.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
Mouse astrocytes

Other Notes

Kannan, K. et al. 1996. Cell Immunol.171, 10.
Rohrer, J., et al. 1996. J. Cell Biol. 132, 565.
Granger, B.L., et al. 1990. J. Biol. Chem. 265, 12036.
Chen, J.W., et al. 1985. J. Cell Biol.101, 85.
Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service