Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

428018

Millipore

Sodium dodecyl sulfate solution

20% in H₂O

Synonym(s):

Sodium n-Dodecyl Sulfate, 20% Solution (w/v), 20% SDS

Sign Into View Organizational & Contract Pricing

Select a Size

200 ML
$148.00

$148.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
200 ML
$148.00

About This Item

Empirical Formula (Hill Notation):
C12H25O4S · Na
CAS Number:
Molecular Weight:
288.38
MDL number:
UNSPSC Code:
12161902
NACRES:
NA.25

$148.00


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

Sodium dodecyl sulfate solution, 20% Solution (w/v), An aqueous solution containing 20% SDS.

Quality Level

description

Merck USA index - 14, 8636

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

clear colorless

shipped in

ambient

storage temp.

15-25°C

SMILES string

[Na+].[S](=O)(=O)([O-])OCCCCCCCCCCCC

InChI

1S/C12H26O4S.Na/c1-2-3-4-5-6-7-8-9-10-11-12-16-17(13,14)15;/h2-12H2,1H3,(H,13,14,15);/q;+1/p-1

InChI key

DBMJMQXJHONAFJ-UHFFFAOYSA-M

General description

An aqueous solution containing 20% SDS.
An ionic detergent useful in the electrophoretic separation of proteins and lipids. Also used in the purification of DNA. Has a CMC of 7-10 mM and a micellar weight of 18,000. Conductivity: 21900-25500 µmhos.

Application


  • Wet SPF: A wet powder suspension for the detection of latent fingermarks on the sticky side of adhesive tape for use in Seychelles.: This study highlights the use of a sodium dodecyl sulfate solution-based wet powder suspension to detect latent fingermarks on adhesive tapes, offering a novel approach in forensic science (Bouzin JT et al., 2024).

  • Neuromorphic sensing of biomolecules covalently immobilised on polydimethyl glutarimide.: The research focuses on using sodium dodecyl sulfate in the process of covalent immobilization of biomolecules, contributing to advancements in neuromorphic sensing technologies (Sylvander LA et al., 2023).

  • Integrated Manufacturing of Suspended and Aligned Nanofibrous Scaffold for Structural Maturation and Synchronous Contraction of HiPSC-Derived Cardiomyocytes.: This paper discusses the role of sodium dodecyl sulfate in the fabrication of nanofibrous scaffolds, crucial for cardiomyocyte maturation in tissue engineering (Liu L et al., 2023).

  • A Sequential Membrane Process of Ultrafiltration Forward Osmosis and Reverse Osmosis for Poultry Slaughterhouse Wastewater Treatment and Reuse.: Sodium dodecyl sulfate is utilized in membrane processes to enhance the treatment and reuse of poultry slaughterhouse wastewater, highlighting its environmental applications (Fatima F et al., 2023).

Warning

Toxicity: Harmful (C)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service