Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

420204-M

Millipore

Histone Lysine Demethylase Inhibitor VII, GSK-J1

Synonym(s):

Histone Lysine Demethylase Inhibitor VII, GSK-J1, JHDM Inhibitor II, 3-(6-(4,5-Dihydro-1H-benzo[d]azepin-3(2H)-yl)-2-(pyridin-2-yl)pyrimidin-4-ylamino)propanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H23N5O2
CAS Number:
Molecular Weight:
389.45
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light yellow

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

SMILES string

N3(CCc4c(cccc4)CC3)c1nc(nc(c1)NCCC(=O)O)c2ncccc2

InChI

1S/C22H23N5O2/c28-21(29)8-12-24-19-15-20(26-22(25-19)18-7-3-4-11-23-18)27-13-9-16-5-1-2-6-17(16)10-14-27/h1-7,11,15H,8-10,12-14H2,(H,28,29)(H,24,25,26)

InChI key

AVZCPICCWKMZDT-UHFFFAOYSA-N

General description

A pyridinyl-pyrimidinylaminopropanoic acid that effectively inhibits KDM6 family H3K27me3 demethylases JMJD3 (IC50 = 0.06 and 18 µM, respectively, by AlphaScreen and MALDI Mass detection method) and UTX (IC50 = 56 µM by MALDI Mass detection) in a α-ketoglutarate-competitive, peptide substrate-non-competitive manner, displaying much reduced potency against JARID1C, JMJD1a, JMJD2a, JMJD2c, JMJD2d, JMJD2e, 112 kinases, as well as a panel of 60 non-kinase enzymes, GPCRs, ion channels, nuclear receptors, and transporters. Recommended for cell-free assays only. For culture treatment, use the cell-permeable pro-drug GSK-J4 (Cat. No. 420205). Structural analysis reveals that the effective inhibition is a result of concerted action of GSK-J1 against the bindings of α-ketoglutarate, histone H3, as well as Fe+2 at the enzyme active site.

Biochem/physiol Actions

Primary Target
H3K27me3 demethylases JMJD3 & UTX

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Heinemann, B., et al. 2014. Nature514 E1.

Kruidenier, L., et al. 2014. Nature514, E2.
Kruidenier, L., et al. 2012. Nature488, 404.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service