Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

420150-M

Millipore

JNK Inhibitor XVI, JNK-IN-8

The JNK Inhibitor XVI, JNK-IN-8 controls the biological activity of JNK. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

JNK Inhibitor XVI, JNK-IN-8, (E)-3-(4-(dimethylamino)but-2-enamido)-N-(3-methyl-4-((4-(pyridin-3-yl)pyrimidin-2-yl)amino)phenyl)benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H29N7O2
CAS Number:
Molecular Weight:
507.59
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Assay

≥97% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light yellow to yellow

solubility

DMSO: 100 mg/mL, clear, yellow

storage temp.

2-8°C

InChI

1S/C29H29N7O2/c1-20-17-24(11-12-25(20)34-29-31-15-13-26(35-29)22-8-5-14-30-19-22)33-28(38)21-7-4-9-23(18-21)32-27(37)10-6-16-36(2)3/h4-15,17-19H,16H2,1-3H3,(H,32,37)(H,33,38)(H,31,34,35)/b10-6+

InChI key

GJFCSAPFHAXMSF-UXBLZVDNSA-N

General description

A cell-permeable, selective, and irreversible type 2 inhibitor of c-Jun N-terminal kinases (JNK) (IC50 = 4.67 nM, 18.7 nM, 980 pM for JNK1, JNK2, and JNK3, respectively). Forms a covalent adduct with the conserved cysteine residue (Cys116 in JNK1) and binds to JNK in an extended type 2 conformation. Blocks the phosphorylation of c-Jun in cells (EC50 = 486 nM in HeLa cells; 338 nM in A375 cells). Exhibits greater cellular specificity for JNK when compared to 40 other kinases screened using a cell-based assay. Shows weaker binding to KIT mutants (IC50 = 92 nM for V559D and 56 nM for T670I) and RIOK2 (120 nM). Does not show any significant inhibitory activity against various other kinases, including ERK, p38, Akt, MSK, and RSK.

Biochem/physiol Actions

Primary Target
JNK1, JNK2, JNK3

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Zhang, T., et al. 2011. Chem. Biol.19, 140

Legal Information

Licensed under pending patent from Dana-Farber Cancer Institute.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service