Skip to Content
MilliporeSigma
All Photos(1)

Documents

420132

Sigma-Aldrich

JAK2 Inhibitor II

The JAK2 Inhibitor II, also referenced under CAS 1837-91-8, controls the biological activity of JAK2. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

JAK2 Inhibitor II, 1,2,3,4,5,6-Hexabromocyclohexane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6Br6
CAS Number:
Molecular Weight:
557.54
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (GC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C6H6Br6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-6H

InChI key

QFQZKISCBJKVHI-UHFFFAOYSA-N

General description

A cell-permeable hexabromocyclohexane compound that interacts with a solvent-accessible pocket near the activation loop of JAK2 and acts as a specific, reversible, and direct inhibitor of JAK2 autophosphorylation (maximal inhibition at 50 µM in BSC-40 cells overexpressing JAK2). Reduces growth hormone-mediated JAK2 activation in γ2A cells that stably express growth hormone receptor and JAK2. Does not block EGFR autophosphorylation in murine fibroblasts.
A cell-permeable, specific, reversible, and direct inhibitor of JAK2 autophosphorylation (maximal inhibition at 50 µM in BSC-40 cells overexpressing JAK2 that interacts with a solvent-accessible pocket near the activation loop of JAK2. Reduces growth hormone-mediated JAK2 activation in γ2A cells that stably express growth hormone receptor and JAK2. Does not block EGFR autophosphorylation in murine fibroblasts.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
JAK2 autophosphorylation
Product does not compete with ATP.
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Sandberg, E.M., et al. 2005. J. Med. Chem.48, 2526.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gerlinde Wernig et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(18), 4757-4762 (2017-04-21)
Fibrotic diseases are not well-understood. They represent a number of different diseases that are characterized by the development of severe organ fibrosis without any obvious cause, such as the devastating diseases idiopathic pulmonary fibrosis (IPF) and scleroderma. These diseases have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service