Recommended Products
Assay
≥95% (reversed phase HPLC)
Quality Level
form
lyophilized
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
desiccated
storage temp.
−20°C
General description
Recombinant rat interferon-γ (IFN-γ) expressed in E. coli as a single, non-glycosylated polypeptide containing 135 amino acids. Purified by chromatographic methods. IFN-γ is a type II cytokine produced by activated T cells and natural killer cells. It plays a significant role immunoregulation and has been implicated in the induction of the expression of class II histocompatibility molecules in cells that do not normally express them, resulting in autoimmune disease.
Biochem/physiol Actions
Reported as U/mg on a lot-specific basis using a cytopathic affect inhibition assay with NIH/3T3 cells challenged with EMC virus.
Warning
Toxicity: Standard Handling (A)
Physical form
Lyophilized from 0.5X PBS, pH 7.2.
Reconstitution
Following reconstitution, refrigerate (4°C) for short-term storage or aliquot and freeze (-20°C) for long-term storage. Stock solutions are stable for up to 7 days at 4°C or for up to 4 months at -20°C. Avoid freeze/thaw cycles of solutions.
Reconstitute with sterile water to ≥100 µg/ml, containing 0.1% HSA or BSA. Further dilute with aqueous buffers just prior to use.
Other Notes
Alvarado, J., et al. 2005. Immunol. Lett.99, 109.
Di Rosa, M., et al. 2005. Clin. Chem. Lab. Med.43, 499.
Malaguarnera, L., et al. 2005. J. Clin. Lab. Anal.19, 128.
Di Rosa, M., et al. 2005. Clin. Chem. Lab. Med.43, 499.
Malaguarnera, L., et al. 2005. J. Clin. Lab. Anal.19, 128.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service