Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

407143-M

Millipore

Anti-IP₃ Receptor Rabbit pAb

liquid, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human, canine, mouse, rat, porcine, bovine

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ITPR1(3708)
mouse ... Itpr1(16438)
rat ... Itpr1(25262)

General description

Rabbit polyclonal antibody purified by ammonium sulfate precipitation and ion-exchange (DEAE) column chromatography. Recognizes the ~260 kDa IP3 receptor protein.
Recognizes all of the ~260 kDa IP3 receptor subtypes.
This Anti-IP₃ Receptor Rabbit pAb is validated for use in ELISA, FC, Immunoblotting, IF, IHC, IP, Radioimmunoassay for the detection of IP₃ Receptor.

Immunogen

a synthetic peptide corresponding to amino acids in the C-terminal cytoplasmic domain of the IP₃ receptor

Application

ELISA (1:2000)

Flow Cytometry (1:1000)

Immunoblotting (1:250)

Immunofluorescence (1:200)

Immunohistochemistry (1:200)

Immunoprecipitation (1:200)

Radioimmunoassay (1:10000)

Warning

Toxicity: Standard Handling (A)

Physical form

In 150 mM NaCl, 10 mM sodium phosphate, pH 7.5.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Bourguignon, L.Y.W., et al. 1994. J. Cell Physiol. 159, 29.
Bourguignon, L.Y.W., et al. 1993. J. Biol. Chem.268, 7290.
Bourguignon, L.Y.W., et al. 1993. Cell Biol. Int.17, 751.
Variables associated with assay conditions will dictate the proper working dilution.



Recommended Protocol for Immunoblotting using anti-IP3 Receptor Antibodies



NOTE: For protein loading, we recommend 1 x 106 cells/gel lane or 100 μg/gel lane.



• TBST

• 20 mM Tris-HCl, pH 7.6

• 150 mM NaCl

• 0.05% Tween®-20 detergent



1. After transfer, block in 15 ml of TBST with 5% dry milk (for polyclonal antibody) or 1% BSA or 1-2% gelatin (for monoclonal antibody).

2. Dilute primary antibody in 5% dry milk (for polyclonal antibody) or 1% BSA or 1-2% gelatin (for monoclonal antibody) in TBST.

3. Incubate with polyclonal antibody for 1 h or with monoclonal antibody for 2 h.

4. Rinse twice rapidly, followed by 1 x 15 min and 2 x 5 min washes with TBST.

5. Dilute secondary antibody also in 5% dry milk (for polyclonal antibody) or 1% BSA or 1-2% gelatin (for monoclonal antibody) in TBST, followed by HRP.

6. Shake for 1 h.

7. Rinse twice rapidly, followed by 1 x 15 min and 2 x 5 min washes with TBST.

8. Mix development solution (1:1) (Amersham ECL kit).

9. In between two pieces of transparency add development solution. Allow to sit for approximately 1 min before exposure to film.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
TWEEN is a registered trademark of Croda International PLC

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service