Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

4071

Sigma-Aldrich

myo-Inositol - CAS 87-89-8 - Calbiochem

Synonym(s):

i-Inositol, meso-Inositol

Sign Into View Organizational & Contract Pricing


About This Item

Assay

98%

form

crystalline solid

color

white

General description

A growth factor for animals as well as microorganisms.
A growth factor for animals as well as microorganisms. The most abundant form of polyols that serves as a structural element of secondary messengers in eukaryotic cells.

Biochem/physiol Actions

A growth factor for animals as well as microorganisms

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J C Ruf et al.
Biochemical medicine and metabolic biology, 48(1), 46-55 (1992-08-01)
Diabetes is known to be associated with an increase in aldose reductase activity, platelet hyperaggregability, lipid peroxidation, and cataract formation. A molecule, D-myo-inositol 1,2,6-trisphosphate (PP-56), derived from phytic acid, could in principle, by supplying myoinositol to tissues and acting as
C P Downes et al.
European journal of biochemistry, 193(1), 1-18 (1990-10-05)
The discovery of the second-messenger functions of inositol 1,4,5-trisphosphate and diacylglycerol, the products of hormone-stimulated inositol phospholipid hydrolysis, marked a turning point in studies of hormone function. This review focuses on the myo-inositol moiety which is involved in an increasingly
B Tesfamariam et al.
The Journal of pharmacology and experimental therapeutics, 263(1), 153-157 (1992-10-01)
A possible relationship between aldose reductase activity and myo-inositol levels and endothelium-dependent relaxations was examined in isolated rabbit aorta incubated with elevated concentrations of glucose (44 mM) for 6 hr to mimic hyperglycemic conditions. Rings of aorta incubated in elevated
B J Holub
Annual review of nutrition, 6, 563-597 (1986-01-01)
Alterations in the level of dietary inositol can significantly influence the concentration of free inositol and inositol-containing phospholipid in the circulation and in selected mammalian tissues and cells. The 1-stearoyl 2-arachidonyl molecular species that commonly predominates in cellular phosphoinositides may

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service