Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

406170

Sigma-Aldrich

IP6K Inhibitor

The IP6K Inhibitor, also referenced under CAS 519178-28-0, controls the biological activity of IP6K.

Synonym(s):

IP6K Inhibitor, N²-(m-Trifluorobenzyl), N⁶-(p-nitrobenzyl)purine, Inositol-1,4,5-trisphosphate 3-Kinase Inhibitor, IP3K Inhibitor, Kcs1 Inhibitor, TNP

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$255.00

$255.00


Please contact Customer Service for Availability


Select a Size

Change View
5 MG
$255.00

About This Item

Empirical Formula (Hill Notation):
C20H16F3N7O2
CAS Number:
Molecular Weight:
443.38
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

$255.00


Please contact Customer Service for Availability

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 10 mg/mL
methanol: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

FC(F)(F)c1cc(ccc1)CNc2nc3[nH]cnc3c(n2)NCc4ccc(cc4)[N+](=O)[O-]

InChI

1S/C20H16F3N7O2/c21-20(22,23)14-3-1-2-13(8-14)10-25-19-28-17(16-18(29-19)27-11-26-16)24-9-12-4-6-15(7-5-12)30(31)32/h1-8,11H,9-10H2,(H3,24,25,26,27,28,29)

InChI key

DDSBPUYZPWNNGH-UHFFFAOYSA-N

General description

A cell-permeable 2,6-disubstituted purine compound that, although originally identified as a reversible, ATP-competitive inhibitor against the conversion of Ins(1,4,5)P3 to Ins(1,3,4,5)P4 by IP3K (IC50 = 18 M; [ATP] = 10 μM), acts in fact as a much more effective inhibitor against the IP6K1/2/3-catalyzed conversion of InsP6 (Cat. No. 407125) to 5-[PP]- InsP6 or InsP7 (IC50 = 0.47 μM against IP6K1; [ATP] = 1 mM) due to a much higher affinity of ATP toward IP3K than IP6K (Km of ATP for the respective enzymes = 0.01 vs 1000 μM). Shown to potently inhibit cellular InsP7 level (IC50 = 0.55 μM in HeLa cultures), while being less potent in increasing the cellular InsP3 accumulation (>10 M in HeLa cultures). TNP is also effective against S. cerevisiae IP6K, Kcs1, but not Vip/PPIP5Ks, enzymes that catalyze the pyrophosphorylation at the 1/3 position of InsP6 and InsP7, in either human or yeast species or two other IP3 metabolizing enzymes, PLC and IP3 5-ppase.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
IP 3-K
Product competes with ATP.
Reversible: yes
Target IC50: 0.47 µM against IP6K1; [ATP] = 1 mM; 18 µM against the conversion of Ins(1,4,5)P3 to Ins(1,3,4,5)P4 by IP3K; [ATP] = 10 µM

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Padmanabhan, U., et al. 2009. J. Biol. Chem.284, 10571.Chang, Y.T., et al. 2002. ChemBioChem3, 897.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service