Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40-301

Millipore

BioTek 50 TS Microplate Washer

The BioTek 50 TS automates the aspirate, dispense shake and soak steps required for many assays including cell-based assays, biomagnetic and polystyrene bead-based assays.

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$13,590.00

$13,590.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
$13,590.00

About This Item

UNSPSC Code:
41100000
NACRES:
NB.13

$13,590.00


Please contact Customer Service for Availability

manufacturer/tradename

Milliplex®

technique(s)

ELISA: suitable
multiplexing: suitable

General description

BioTek′s 50 TS Microplate Washer is a compact microplate washing system with functionality that is unsurpassed in its class. The color touchscreen provides a visual interface with menu-driven programming that makes creating protocols fast and intuitive. Its performance for conventional ELISA plate washing is excellent, but the 50 TS offers much more. Its modularity makes it ideal for cellbased assay washing, biomagnetic separation and vacuum filtration processes.

The 50 TS is an affordable choice for automating the wash steps of a variety of applications in clinical and research laboratories. Used in conjunction with the 800 TS Absorbance Reader or other detection system, the 50 TS offers a welcome upgrade from manual processing - bringing convenience and consistently high quality results to your laboratory′s plate washing workflows.

Features
  • Application versatility: ELISA, cell-based assays and bead-based assays--Applications for the 50 TS extend far beyond simple dispense and aspirate routines typical of many ELISA processes. Optimize fluid delivery for gentle cell-based assay washing, or run bead-based processes with the available biomagnetic and vacuum filtration modules.
  • Color touchscreen makes programming quick and easy--The 50 TS software includes pre-defined protocols with commonly used wash parameters for quick selection. Creating custom protocols onboard the 50 TS is easy – the touchscreen and menu-driven software make multi-step program creation intuitive and simple!
  • Automated switching of up to 3 buffers for even greater automation--To facilitate complex wash protocols or automated maintenance routines, the automated buffer switching capability adds even greater automation to the system.
  • Automated, built-in maintenance routines for continued reliable operation--Pre-defined, automated maintenance routines, including daily and overnight protocols keep the fluid path clean and prevent buildup of salt, protein or other material that can block manifold tubes, causing inadequate washing.


General Specifications
Microplate types: 24-, 96-, 384-well plates (96- and 384- well: standard and low profile plates; solid and filter bottom (option), 24-well plates with 4-well manifold)
Onboard software: Up to 75 user-programmable protocols; Quick menu; Create or edit custom protocols; Run protocols created onboard or downloaded from LHC Software
Separation: Biomagnetic separation (“M” configurations)
Shaking: Programmable in minutes and seconds up to 30 minutes; 5 intensities from 15-19 Hz
Soaking: Programmable in minutes and seconds up to 30 minutes
User interface: 4.3" color LCD touchscreen display

Physical Characteristics
Power External 24VDC power supply compatible with 100-240VAC @ 50-60Hz
Dimensions 15 x 15 x 8 inches (35.6 x 40.6 x 16.5 cm)
Weight 22 lbs (9.8 kg)
Connectivity 1 USB port for computer control

Application

The BioTek 50 TS automates the aspirate, dispense shake and soak steps required for many assays including cell-based assays, biomagnetic and polystyrene bead-based assays.
For use with MILLIPLEX MAP assays and ELISA. See product protocol for specific use instructions.

Legal Information

MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service