Skip to Content
MilliporeSigma
All Photos(1)

Documents

401488

Sigma-Aldrich

IKK-3 Inhibitor IX

The IKK-3 Inhibitor IX, also referenced under CAS 862812-98-4, controls the biological activity of IKK-3. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.

Synonym(s):

IKK-3 Inhibitor IX, 5-(5,6-Dimethoxybenzimidazol-1-yl)-3-(2-methanesulfonyl-benzyloxy)-thiophene-2-carbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H19N3O5S2
CAS Number:
Molecular Weight:
469.53
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

beige

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C22H19N3O5S2/c1-28-17-8-15-16(9-18(17)29-2)25(13-24-15)22-10-19(20(11-23)31-22)30-12-14-6-4-5-7-21(14)32(3,26)27/h4-10,13H,12H2,1-3H3

InChI key

LDTAHRLHGHFHKP-UHFFFAOYSA-N

General description

A thiophenecarbonitrile compound that acts as a potent ATP-competitive inhibitor of IKK-3 (IKK-ε, IC50 = 40 nM) with ~20-fold selectivity over PLK1 and more than 125-fold selectivity over a panel of 9 other commonly studied kinases, including IKK-1/2 (IKK-α/β). Reported to be inactive against cytochrome P450 isozymes (IC50<50 µM) 1A2, 2D6, 2C9, and 3A4.

Biochem/physiol Actions

Cell permeable: no
Primary Target
IKK-3
Product competes with ATP.
Reversible: no
Target IC50: 40 nM against IKK-3 (IKK-ε)

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Bamborough, P., et al. 2006. Bioorg. Med. Chem. Lett.16, 6236.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service