Skip to Content
MilliporeSigma
All Photos(1)

Documents

401315-M

Millipore

Goat Anti-Rabbit IgG, H & L Chain Specific Peroxidase Conjugate

liquid, Calbiochem®

Synonym(s):

Goat IgG antibody, Goat anti-Rabbit IgG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

goat

Quality Level

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

liquid

contains

0.1% Kathon as preservative

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

isotype

IgG

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Immunoaffinity purified goat polyclonal antibody conjugated to horseradish peroxidase (HRP). Recognizes rabbit IgG, heavy and light chains.
This Goat Anti-Rabbit IgG, H & L Chain Specific Peroxidase Conjugate is validated for use in Enzyme Immunoassay, Immunoelectrophoresis for the detection of Rabbit IgG, H & L Chain Specific.

Application

Enzyme Immunoassay (1:10,000)

Immunoelectrophoresis (see comments)

Warning

Toxicity: Standard Handling (A)

Physical form

In 10 mM PBS, 0.5 mM EDTA, 10% mannose, 2% sucrose, 1% BSA, pH 7.6.

Other Notes

o-Phenylenediamine, 1 mg/ml in citrate buffer, pH 4.5, 0.1% urea peroxide at 25°C, was used for testing immunoenzyme reactivity. Monospecific for rabbit IgG, heavy and light chains, as determined by immunoelectrophoresis against normal rabbit serum. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service