Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

399698

Millipore

5-Hydroxy-L-tryptophan

Serotonin precursor.

Synonym(s):

5-Hydroxy-L-tryptophan, L-5-HTP

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$682.00

$682.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$682.00

About This Item

Empirical Formula (Hill Notation):
C11H12N2O3
CAS Number:
Molecular Weight:
220.22
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.21

$682.00


Please contact Customer Service for Availability

Request a Bulk Order

description

Merck USA index - 14, 4847

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white to off-white

solubility

acidic solutions: soluble
water: soluble

shipped in

ambient

storage temp.

2-8°C

SMILES string

[N+H3][C@@H](Cc1c2c([nH]c1)ccc(c2)O)C(=O)[O-]

InChI

1S/C11H12N2O3/c12-9(11(15)16)3-6-5-13-10-2-1-7(14)4-8(6)10/h1-2,4-5,9,13-14H,3,12H2,(H,15,16)/t9-/m0/s1

InChI key

LDCYZAJDBXYCGN-VIFPVBQESA-N

General description

Serotonin precursor.

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution in acidic solutions, this product will remain stable for up to one week. It is recommended that aqueous stock solutions be prepared just prior to use.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service