391336-M
BICINE
ULTROL® Grade, >99% (titration), zwitterionic amino acid buffer useful pH range 7.5-9.0
Synonym(s):
Bicine, ULTROL® Grade, N,N- bis-(2-Hydroxyethyl)glycine, N,N-bis-(2-Hydroxyethyl)glycine
About This Item
Recommended Products
Quality Level
Assay
>99% (titration)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
color
white
pKa (20 °C)
8.4
solubility
water: 5 mg/mL
anion traces
sulfate (SO42-): ≤0.005%
cation traces
Na: ≤0.005%
heavy metals (as Pb): <0.0005%
storage temp.
10-30°C
InChI
1S/C6H13NO4.Na/c8-3-1-7(2-4-9)5-6(10)11;/h8-9H,1-5H2,(H,10,11);/q;+1/p-1
InChI key
MFBDBXAVPLFMNJ-UHFFFAOYSA-M
General description
Warning
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service