Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

382175-M

Millipore

Histone Deacetylase Inhibitor VIII

The Histone Deacetylase Inhibitor VIII, also referenced under CAS 1045792-66-2, controls the biological activity of Histone Deacetylase. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

Histone Deacetylase Inhibitor VIII, N-(4-(3-(((7-Hydroxyamino-7-oxoheptyl)amino)carbonyl)-5-isoxazolyl)phenyl)-1,1-dimethylethylester, carbamic acid, HDAC Inhibitor VIII

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H30N4O6
CAS Number:
Molecular Weight:
446.50
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

beige to light brown

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

SMILES string

N(O)C(=O)CCCCCCNC(=O)c1n[o]c(c1)c2ccc(cc2)NC(=O)OC(C)(C)C

InChI

1S/C22H30N4O6/c1-22(2,3)31-21(29)24-16-11-9-15(10-12-16)18-14-17(26-32-18)20(28)23-13-7-5-4-6-8-19(27)25-30/h9-12,14,30H,4-8,13H2,1-3H3,(H,23,28)(H,24,29)(H,25,27)

InChI key

WWGBHDIHIVGYLZ-UHFFFAOYSA-N

General description

A cell-permeable, active site Zn2+-targeting phenylisoxazolo-hydroxamate that acts a potent and highly selective HDAC6 inhibitor (IC50 = 0.002, 0.42, 90.7, 252, 271 and 6851 nM against HDAC-6, -3, -10, -2, -1 and -8, respectively). Shown to be more potent than SAHA in inhibiting the proliferation of a panel of pancreatic cancer cell lines (IC50<1.0 µM).

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Kozikowski, A.P., et al. 2008. J. Med. Chem.51, 4370.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service