Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

382173

Sigma-Aldrich

Histone Deacetylase Inhibitor VII, 106

The Histone Deacetylase Inhibitor VII, 106, also referenced under CAS 937039-45-7, controls the biological activity of Histone Deacetylase. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

Histone Deacetylase Inhibitor VII, 106, N¹-(2-Aminophenyl)-N⁷- p-tolylheptanediamide, Pimelic Diphenylamide 106

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$628.00

$628.00


Please contact Customer Service for Availability


Select a Size

Change View
5 MG
$628.00

About This Item

Empirical Formula (Hill Notation):
C20H25N3O2
CAS Number:
Molecular Weight:
339.43
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$628.00


Please contact Customer Service for Availability

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 10 mg/mL
ethanol: 2.5 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

N(c2c(cccc2)N)C(=O)CCCCCC(=O)Nc1ccc(cc1)C

InChI

1S/C20H25N3O2/c1-15-11-13-16(14-12-15)22-19(24)9-3-2-4-10-20(25)23-18-8-6-5-7-17(18)21/h5-8,11-14H,2-4,9-10,21H2,1H3,(H,22,24)(H,23,25)

InChI key

WTKBRPXPNAKVEQ-UHFFFAOYSA-N

General description

A cell-permeable p-tolylbenzamide and a Histone Deacetylase (HDAC) Inhibitor IV (Cat. No. 382170) analog that acts as a selective inhibitor against class I HDAC1,2,3 (IC50 = 0.15, 0.76, and 0.37 µM with 15, 30, and 180 min preincubation, respectively), while exhibiting much lower acitivity against class I HDAC8 (IC50 ≥ 5µM with 3 h preincubation) and no activity against class II HDAC4/5/7 even at concentrations as high as 180µM. Although the mode of inhibition is mechanistically competitive and reversible, due to the slow- and tight-binding nature, long preincubation is often required for effective in vitro enzyme inhibition, while 106-induced cellular H3 hyperacetylation is shown to persist for more than 6 hours after inhibitor removal by washing in GM15850 culture. 106 is reported to be less cytotoxic than TSA (Cat. No. 647925), MS-275, and SAHA (EC50 = 6.3, 0.328, 0.768, and 1.5 µM, respectively, in GM15850 killing).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Chou, C.J., et al. 2008. J. Biol. Chem.283, 35402.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service