Skip to Content
MilliporeSigma
All Photos(1)

Documents

3779

Millipore

Histamine, Free Base

Synonym(s):

Histamine, Free Base

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9N3
CAS Number:
Molecular Weight:
111.15
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.77

Assay

≥97% (TLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

water: 1 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C5H9N3/c6-2-1-5-3-7-4-8-5/h3-4H,1-2,6H2,(H,7,8)

InChI key

NTYJJOPFIAHURM-UHFFFAOYSA-N

General description

A potent vasodilator found in normal tissues and blood. Activates nitric oxide synthase.
A potent vasodilator found in normal tissues and blood. Activates nitric oxide synthase. Formed by the decarboxylation of L-histidine. Activation of histamine H1 receptors leads to Ca2+ mobilization, whereas histamine H2 receptor activation stimulates adenylate cyclase activity in neurons. Stimulated time- and concentration-dependent increase in p38 and p42/p44 MAP kinases in DDT(1) MF2 cells.
A potent vasodilator found in normal tissues and blood. Formed by the decarboxylation of L-histidine. Activates nitric oxide synthase (NOS), resulting in the release of nitric oxide. Activation of histamine H1 receptors leads to Ca2+ mobilization; whereas, histamine H2 receptor activation stimulates adenylate cyclase activity in neurons. Inhibits the synthesis of IL-2 and γ-IFN in peripheral blood mononuclear cells and lipopolysaccharide-induced synthesis of TNF-α in monocytes via H2 receptor activation.

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Robinson, A.J., and Dickerson, J.M. 2001. Br. J. Pharmacol.133, 178.
Izumi, H., et al. 1995. Am. J. Obstet. Gynecol.172, 1477.
Vannier, E., and Dinarello, C.A. 1994. J. Biol. Chem.269, 9952.
Yan, Q.Z., et al. 1994. Agents Actions 41, C111.
Moncada, S., et al. 1991. Pharmacol. Rev. 43, 109.
Bloom, F.E. 1990. in Pharmacological Basis of Therapeutics p. 224 (Gilman, A.G., et al., Eds.) Pergamon Press, New York.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service