Skip to Content
MilliporeSigma
All Photos(1)

Documents

371970

Sigma-Aldrich

HA 1077, Dihydrochloride

A cell-permeable, reversible, and ATP-competitive Ca2+ antagonist with anti-vasospastic properties.

Synonym(s):

HA 1077, Dihydrochloride, Fasudil, (5-Isoquinolinesulfonyl)homopiperazine, 2HCl, PKG Inhibitor VII

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H17N3O2S · 2HCl
CAS Number:
Molecular Weight:
364.29
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 1 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C14H17N3O2S/c18-20(19,17-9-2-6-15-8-10-17)14-4-1-3-12-11-16-7-5-13(12)14/h1,3-5,7,11,15H,2,6,8-10H2

InChI key

NGOGFTYYXHNFQH-UHFFFAOYSA-N

General description

A cell-permeable, reversible, and ATP-competitive Ca2+ antagonist with anti-vasospastic properties. Inhibits protein kinase A (Ki = 1.6 µM), protein kinase G (Ki = 1.6 µM), and myosin light chain kinase (Ki = 3.6 µM). Also reported to potently inhibit Rho-associated kinase (ROCK; IC50 = 10.7 µM). Prevents apoptosis and enhances the survival and cloning efficiency of dissociated hES cells without affecting their pluripotency.
A cell-permeable, reversible, and ATP-competitive Ca2+ antagonist with anti-vasospastic properties. Inhibits protein kinase A (Ki = 1.6 µM), protein kinase G (Ki = 1.6 µM), and myosin light chain kinase (Ki = 36 µM). Also reported to potently inhibit Rho-associated kinase (ROCK; IC50 = 10.7 µM). Prevents apoptosis and enhances the survival and cloning efficiency of dissociated hES cells without affecting their pluripotency.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PKA
Product competes with ATP.
Reversible: yes
Target IC50: 10.7 µM inhibiting Rho-associated kinase (ROCK)
Target Ki: .6 µM, 1.6 µM, 36 µM, inhibiting protein kinase A, protein kinase G, myosin light chain kinase, respectively

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, refrigerate (4°C). Aqueous stock solutions are stable for up to 6 months at 4°C.

Other Notes

Watanabe, K., et al. 2007. Nature Biotech.25, 681.
Swärd, K., et al. 2000. J. Physiol.522, 33.
Takizawa, S., et al. 1993. Eur. J. Pharmacol. 250, 431.
Seto, M., et al. 1991. Eur. J. Pharmacol.195, 267.
Shirotani, M., et al. 1991. J. Pharmacol. Exp. Ther.259, 738.
Asano, T., et al. 1990. Eur. J. Pharmacol.190, 365.
Sasaki, Y., and Sasaki, Y. 1990. Biochem. Biophys. Res. Commun.171, 1182.
Asano, T., et al. 1989. Br. J. Pharmacol. 98,1091.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service