Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

371806

Sigma-Aldrich

GTP-14564

A cell-permeable, reversible, and ATP-competitive tricyclic benzofurano-indazolo compound that acts as a potent and specific inhibitor of class III receptor tyrosine kinases.

Synonym(s):

GTP-14564, 1-Phenyl-3-H-8-oxa-2,3-diaza-cyclopenta[a]inden, 3-Phenyl-1H-benzofuro[3,2-c]pyrazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10N2O
CAS Number:
Molecular Weight:
234.25
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C15H10N2O/c1-2-6-10(7-3-1)13-15-14(17-16-13)11-8-4-5-9-12(11)18-15/h1-9H,(H,16,17)

InChI key

DZQLVVLATXPWBK-UHFFFAOYSA-N

General description

A cell-permeable, reversible, ATP-competitive, and tricyclic, benzofurano-indazolo compound that acts as a potent and specific inhibitor of class III receptor tyrosine kinases (IC50 = 300 nM for c-fms, c-kit, wt-FLT3, and internal tandem duplication (ITD)-FLT3; 1 µM for PDGFRβ). Does not affect the activities of KDR, EGFR, HER2, Abl, Src, PKA, Akt, PKC, MEK, or ERK1/2 (IC50 >10 µM). Inhibits FLT-3 ligand-dependent growth of Ba/F3 cells expressing wild type FLT-3. Reported to be useful in differentiating signal transduction pathways activated by wt-FLT3 and ITD-FLT3 in Ba/F3 cells.
A cell-permeable, reversible, and ATP-competitive tricyclic benzofurano-indazolo compound that acts as a potent and specific inhibitor of class III receptor tyrosine kinases (IC50 = 0.3 µM for c-fms, c-kit, wt-FLT3 and ITD-FLT3; 1.0 µM for PDGFRβ). Does not affect the activities of KDR, EGFR, HER2, Abl, Src, PKA, AKT, PKC, MEK, or ERK1/2 (IC50 ≥ 10 µM). Has been used successfully to differentiate between the signalling pathways activated by wt-FLT3 and ITD-FLT3 in Ba/F3 cells.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Class III receptor tyrosine kinases
Product competes with ATP.
Reversible: yes
Target IC50: 0.3 µM for c-fms, c-kit, wt-FLT3 and ITD-FLT3; 1.0 µM for PDGFRβ

Packaging

Packaged under inert gas

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Other Notes

Murata, K., et al. 2003. J. Biol. Chem.278, 32892.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service